2,4-Difluoro-6-nitroaniline ≥98%

Supplier: Matrix Scientific

005124-5G 005124-25G
101788-292EA 21.54 USD
101788-292 101788-294
2,4-Difluoro-6-nitroaniline ≥98%
2,4-Difluoro-6-nitroaniline

Hazard code(s): Irritant

Formula: C₆H₄F₂N₂O₂
MW: 174.11 g/mol
MDL Number: MFCD00034058
CAS Number: 364-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR