1,3-Dichloro-4-fluoro-6-nitrobenzene

Supplier: Matrix Scientific

Synonyms: 1,5-Dichloro-2-fluoro-4-nitrobenzene, 2,4-Dichloro-5-fluoronitrobenzene

005112-1G 005112-5G
101788-268EA 63.08 USD
101788-268 101788-270
1,3-Dichloro-4-fluoro-6-nitrobenzene
1,3-Dichloro-4-fluoro-6-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₂Cl₂FNO₂
MW: 209.99 g/mol
Boiling Pt: 56 °C (0.5mm)
MDL Number: MFCD00115003
CAS Number: 2105-59-1

Order Now

Specification Test Results

Boiling point 56°C/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR