2,3-Difluoro-p-toluic acid

Supplier: Matrix Scientific

005073-1G 005073-5G
101788-228EA 18.46 USD
101788-228 101788-230
2,3-Difluoro-p-toluic acid
2,3-Difluoro-p-toluic acid
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD01631643
CAS Number: 261763-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR