2,3-Difluoro-4-methoxyphenol

Supplier: Matrix Scientific

Synonyms: 2,3-Difluor-4-hydroxyanisole

005068-1G 005068-5G
101788-208EA 60 USD
101788-208 101788-210
2,3-Difluoro-4-methoxyphenol
2,3-Difluoro-4-methoxyphenol
Formula: C₇H₆F₂O₂
MDL Number: MFCD01631505
CAS Number: 261763-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR