3,5-Dichloro-4-iodobenzotrifluoride ≥97%

Supplier: Matrix Scientific

005067-1G 005067-5G
101788-204EA 70.77 USD
101788-204 101788-206
3,5-Dichloro-4-iodobenzotrifluoride ≥97%
3,5-Dichloro-4-iodobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₂Cl₂F₃I
MW: 340.9 g/mol
Boiling Pt: 80…82 °C (1.2mm)
MDL Number: MFCD00084997
CAS Number: 175205-56-8

Order Now

Specification Test Results

Boiling point 80-82°C/1.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR