3,5-Dichloro-4-fluorobenzotrifluoride ≥98%

Supplier: Matrix Scientific

Synonyms: 1,3-Dichloro-2-fluoro-5-(trifluoromethyl)benzene, 3,5-Dichloro-4-fluorobenzotrifluoride

005062-1G 005062-5G
101788-196EA 38.46 USD
101788-196 101788-198
3,5-Dichloro-4-fluorobenzotrifluoride ≥98%
3,5-Dichloro-4-fluorobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₂Cl₂F₄
MW: 232.99 g/mol
Boiling Pt: 137…140 °C
Density: 1.55 g/cm³ (20 °C)
MDL Number: MFCD00068183
CAS Number: 77227-81-7

Order Now

Specification Test Results

Boiling point 137-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR