2,6-Difluoro-3-methylbenzaldehyde

Supplier: Matrix Scientific

Synonyms: 2,6-Difluoro-3-tolualdehyde, 2,6-Difluoro-m-tolualdehyde, 2,6-Difluoro-m-tolylaldehyde, 2,6-Difluoro-3-tolylaldehyde

005013-1G 005013-5G
101788-156EA 32.31 USD
101788-156 101788-158
2,6-Difluoro-3-methylbenzaldehyde
2,6-Difluoro-3-methylbenzaldehyde
Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD01631333
CAS Number: 261763-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR