1,2-Dichloro-1,1,2-trifluoro-2-iodoethane ≥97%

Supplier: Matrix Scientific

004971-5G 004971-25G
101788-098EA 84.62 USD
101788-098 101788-100
1,2-Dichloro-1,1,2-trifluoro-2-iodoethane ≥97%
1,2-Dichloro-1,1,2-trifluoro-2-iodoethane

Hazard code(s): Irritant, Light Sensitive

Formula: ClF₂CCClFI
MW: 278.83 g/mol
Boiling Pt: 100 °C
MDL Number: MFCD00039402
CAS Number: 354-61-0
EINECS: 206-565-1

Order Now

Specification Test Results

Boiling point 100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR