3-(3-Bromophenyl)propionic acid ≥98%

Supplier: Matrix Scientific

004940-5G 004940-25G
101788-064EA 115.38 USD
101788-064 101788-066
3-(3-Bromophenyl)propionic acid ≥98%
3-(3-Bromophenyl)propionic acid

Hazard code(s): Irritant

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD01310792
CAS Number: 42287-90-1

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR