3-(4-Aminophenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Aminohydrocinnamic acid, 3-(4-Aminophenyl)propanoic acid

004937-1G 004937-5G 004937-25G
101788-050EA 43.08 USD
101788-050 101788-052 101788-054
3-(4-Aminophenyl)propionic acid ≥98%
3-(4-Aminophenyl)propionic acid

Hazard code(s): Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 135…137 °C
MDL Number: MFCD00017118
CAS Number: 2393-17-1
EINECS: 219-245-1

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR