1-(4-Bromophenyl)imidazole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(4-Bromophenyl)-1H-imidazole

004932-1G 004932-5G
101788-030EA 50.77 USD
101788-030 101788-032
1-(4-Bromophenyl)imidazole ≥98%
1-(4-Bromophenyl)imidazole

Hazard code(s): Irritant

Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD00060489
CAS Number: 10040-96-7

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR