N-(6-Bromohexyl)phthalimide ≥98%

Supplier: Matrix Scientific

004882-1G 004882-5G
101787-952EA 38.46 USD
101787-952 101787-954
N-(6-Bromohexyl)phthalimide ≥98%
N-(6-Bromohexyl)phthalimide

Hazard code(s): Irritant

Formula: C₁₄H₁₆BrNO₂
MW: 310.19 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD00023098
CAS Number: 24566-79-8

Order Now

Specification Test Results

Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR