Perfluorooctane ≥99%

Supplier: Matrix Scientific

Synonyms: Octadecafluorooctane

004851-5G 004851-25G
101787-904EA 15.38 USD
101787-904 101787-906
Perfluorooctane ≥99%
Perfluorooctane

Hazard code(s): Irritant

Formula: C₈F₁₈
MW: 438.06 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: -25 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00042083
CAS Number: 307-34-6
EINECS: 206-199-2

Order Now

Specification Test Results

Boiling point 99-100°C
Density 1.73
ND20 1.282

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR