Decafluorobenzophenone ≥98%

Supplier: Matrix Scientific

Synonyms: Decafluorobenzophenone, Perfluorobenzophenone

004837-1G 004837-5G
101787-882EA 127.69 USD
101787-882 101787-884
Decafluorobenzophenone ≥98%
Decafluorobenzophenone

Hazard code(s): Irritant

Formula: C₁₃F₁₀O
MW: 362.13 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000295
CAS Number: 853-39-4
EINECS: 212-717-8

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR