1,3-Diiodobenzene ≥98%

Supplier: Matrix Scientific
Warning

004832-25G 004832-100G
101787-870EA 49.23 USD
101787-870 101787-872
1,3-Diiodobenzene ≥98%
1,3-Diiodobenzene

Hazard code(s): Irritant

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2.76 g/cm³ (0 °C)
MDL Number: MFCD00041731
CAS Number: 626-00-6
EINECS: 210-921-1

Order Now

Specification Test Results

Melting point 34-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR