3-Chloro-o-xylene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2,3-dimethylbenzene, 3-Chloro-1,2-dimethylbenzene

004816-25G 004816-100G
101787-856EA 61.54 USD
101787-856 101787-858
3-Chloro-o-xylene ≥98%
3-Chloro-o-xylene

Hazard code(s): Irritant

Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 78 °C (22mm)
Density: 1.06 g/cm³
MDL Number: MFCD00060689
CAS Number: 608-23-1

Order Now

Specification Test Results

Boiling point 78°C/22mm
Density 1.06

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR