4-Bromobenzhydrol ≥98%

Supplier: Matrix Scientific

Synonyms: p-Bromobenzhydrol, 4-Bromophenyl phenyl carbinol, 4-Bromodiphenylmethanol, 4-Bromo-α-phenylbenzyl alcohol, 4-Bromobenzhydryl alcohol, (4-Bromophenyl)(phenyl)methanol

004731-1G 004731-5G
101787-750EA 21.54 USD
101787-750 101787-752
4-Bromobenzhydrol ≥98%
4-Bromobenzhydrol

Hazard code(s): Irritant

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD00016851
CAS Number: 29334-16-5
EINECS: 249-568-3

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR