4-Iodophenylboronic acid

Supplier: Matrix Scientific

Synonyms: (4-Iodophenyl)boronic acid, Dihydroxy(4-iodophenyl)borane, 4-Iodobenzeneboronic acid, (4-Iodophenyl)metaboric acid

004719-1G 004719-5G
101787-736EA 29.23 USD
101787-736 101787-738
4-Iodophenylboronic acid
4-Iodophenylboronic acid
Formula: C₆H₆BIO₂
MW: 247.83 g/mol
MDL Number: MFCD01319014
CAS Number: 5122-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR