(4-Iodophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-Iodophenyl)acetonitrile, 4-Iodobenzyl cyanide, 4-Iodophenylacetonitrile

004717-1G 004717-5G 004717-25G
101787-726EA 52.31 USD
101787-726 101787-728 101787-730
(4-Iodophenyl)acetonitrile ≥97%
(4-Iodophenyl)acetonitrile

Hazard code(s): Irritant, Irritant-Harmful, Light Sensitive

Formula: C₈H₆IN
MW: 243.05 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 56 - 57 °C
MDL Number: MFCD00060306
CAS Number: 51628-12-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 152°C/ 3mm
Melting point 55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR