α,α,α-Trifluoro-2,5-diaminotoluene ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 2,5-Diaminobenzotrifluoride, 2-(Trifluoromethyl)benzene-1,4-diamine, 2-(Trifluoromethyl)-p-phenylenediamine, 2-(Trifluoromethyl)-1,4-phenylenediamine

004678-1G 004678-5G
101787-672EA 47.69 USD
101787-672 101787-674
α,α,α-Trifluoro-2,5-diaminotoluene ≥97% for synthesis
α,α,α-Trifluoro-2,5-diaminotoluene

Hazard code(s): Irritant-Harmful

Formula: C₇H₇F₃N₂
Boiling Pt: 117 - 118 °C (7 -rr)
Melting Pt: 58 °C
MDL Number: MFCD00036101
CAS Number: 364-13-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 57-59°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR