α,α,α-Trifluoro-3,4-diaminotoluene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)-1,2-phenylenediamine, 4-(Trifluoromethyl)benzene-1,2-diamine, 3,4-Diaminobenzotrifluoride, 4-(Trifluoromethyl)-o-phenylenediamine

004676-1G 004676-5G
101787-668EA 53.85 USD
101787-668 101787-670
α,α,α-Trifluoro-3,4-diaminotoluene ≥95%
α,α,α-Trifluoro-3,4-diaminotoluene

Hazard code(s): Toxic, Irritant-Harmful

Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Boiling Pt: 101 - 102 °C (7 -rr)
Melting Pt: 58 °C
MDL Number: MFCD00042456
CAS Number: 368-71-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR