1,2-Diamino-3,4-difluorobenzene ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3,4-Difluoro-1,2-phenylenediamine, 3,4-Difluorobenzene-1,2-diamine, 3,4-Difluoro-o-phenylenediamine

004667-1G 004667-5G
101787-646EA 44.62 USD
101787-646 101787-648
1,2-Diamino-3,4-difluorobenzene ≥98% for synthesis
1,2-Diamino-3,4-difluorobenzene

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₆F₂N₂
Melting Pt: 58…60 °C
MDL Number: MFCD00153128
CAS Number: 153505-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR