2-Iodo-5-nitrotoluene ≥99%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-Iodo-2-methyl-4-nitrobenzene
004657-5G
004657-25G
101787-630EA
20
USD
101787-630
101787-632
2-Iodo-5-nitrotoluene ≥99%
2-Iodo-5-nitrotoluene
Hazard code(s): Irritant
Formula:
C₇H₆INO₂ MW: 263.03 g/mol Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051517 CAS Number: 5326-38-5 EINECS: 226-204-1 |
Specification Test Results
Melting point | 99-101°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...