2-Iodo-5-nitroanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Iodo-2-methoxy-4-nitrobenzene

004651-25G 004651-100G
101787-618EA 61.54 USD
101787-618 101787-620
2-Iodo-5-nitroanisole ≥98%
2-Iodo-5-nitroanisole

Hazard code(s): Irritant

Formula: C₇H₆INO₃
MW: 279.03 g/mol
Melting Pt: 133…135 °C
MDL Number: MFCD00024590
CAS Number: 5458-84-4

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR