3-Benzylaniline ≥98%

Supplier: Matrix Scientific

004637-1G 004637-5G
101787-610EA 69.23 USD
101787-610 101787-612
3-Benzylaniline ≥98%
3-Benzylaniline

Hazard code(s): Irritant

Formula: C₁₃H₁₃N
MW: 183.25 g/mol
Boiling Pt: 126 °C (2mm)
Melting Pt: 48…52 °C
MDL Number: MFCD01310782
CAS Number: 61424-26-8

Order Now

Specification Test Results

Boiling point 126°C/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR