Cyclobutanecarboxylic acid ≥98%
Supplier: Matrix Scientific
|
Danger
|
004628-25G
004628-100G
101787-602EA
40
USD
101787-602
101787-604
Cyclobutanecarboxylic acid ≥98%
Cyclobutanecarboxylic acid
Hazard code(s): Stench
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 195 °C (1013 hPa) Melting Pt: -20 °C Density: 1.06 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001323 CAS Number: 3721-95-7 EINECS: 223-072-7 UN: 3265 ADR: 8,III |
Specification Test Results
Boiling point | 195°C |
Melting point | -20 to -7. |
Density | 1.047 |
ND20 | 1.4433 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...