2-Thiophenecarbonitrile ≥97%
Supplier: Matrix Scientific
004625-25G
004625-100G
101787-590EA
33.85
USD
101787-590
101787-592
2-Thiophenecarbonitrile ≥97%
2-Thiophenecarbonitrile
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₅H₃NS Boiling Pt: 190 - 192 °C (1013 hPa) Density: 1.18 g/cm³ (15 °C) |
MDL Number:
MFCD00005416 CAS Number: 1003-31-2 EINECS: 213-706-0 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 191-193°C |
Density | 1.172 |
ND20 | 1.563 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...