1-Naphthyl boronic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

004614-25G 004614-100G
101787-578EA 40 USD
101787-578 101787-580
1-Naphthyl boronic acid ≥97%
1-Naphthyl boronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Melting Pt: 259 °C
MDL Number: MFCD00019722
CAS Number: 13922-41-3

Order Now

Specification Test Results

Melting point 210-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR