2,5-Dibromothiophene ≥95%
Supplier: Matrix Scientific
|
Warning
|
004596-100G
004596-250G
101787-558EA
84.62
USD
101787-558
101787-560
2,5-Dibromothiophene ≥95%
2,5-Dibromothiophene
Hazard code(s): Irritant
Formula:
C₄H₂Br₂S Boiling Pt: 211 °C Melting Pt: –6 °C Density: 2.147 g/cm³ |
MDL Number:
MFCD00005420 CAS Number: 3141-27-3 EINECS: 221-547-3 UN: 2810 ADR: 6.1,II |
Specification Test Results
Boiling point | 211°C |
Melting point | -6°C |
Density | 2.147 |
ND20 | 1.629 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...