2,5-Dibromophenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

004587-1G 004587-5G
101787-534EA 69.23 USD
101787-534 101787-536
2,5-Dibromophenyl isothiocyanate ≥98%
2,5-Dibromophenyl isothiocyanate

Hazard code(s): Irritant, Lachrymator, Refrigerate

Formula: C₇H₃Br₂NS
Melting Pt: 49…51 °C
MDL Number: MFCD00041041
CAS Number: 98041-67-9

Order Now

Specification Test Results

Melting point 49-51°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR