1,4-Dibromooctafluorobutane ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1,4-Dibromoperfluorobutane

004586-5G 004586-25G
101787-530EA 118.46 USD
101787-530 101787-532
1,4-Dibromooctafluorobutane ≥98% for synthesis
1,4-Dibromooctafluorobutane

Hazard code(s): Irritant

Formula: C₄Br₂F₈
MW: 359.84 g/mol
Boiling Pt: 97 °C
MDL Number: MFCD00153112
CAS Number: 335-48-8

Order Now

Specification Test Results

Boiling point 97°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR