1,8-Dibromohexadecafluorooctane ≥96%

Supplier: Matrix Scientific

004585-5G 004585-25G
101787-526EA 146.15 USD
101787-526 101787-528
1,8-Dibromohexadecafluorooctane ≥96%
1,8-Dibromohexadecafluorooctane

Hazard code(s): Irritant

Formula: C₈Br₂F₁₆
Boiling Pt: 180 °C
Melting Pt: 40 °C
MDL Number: MFCD00153111
CAS Number: 812-58-8

Order Now

Specification Test Results

Boiling point 180°C
Melting point 40°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR