2,7-Dibromofluorene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2,7-Dibromo-9H-fluorene

004577-5G 004577-25G 004577-100G
101787-514EA 15.38 USD
101787-514 101787-516 101787-518
2,7-Dibromofluorene ≥98%
2,7-Dibromofluorene

Hazard code(s): Irritant

Formula: C₁₃H₈Br₂
MW: 324.01 g/mol
Melting Pt: 164…166 °C
MDL Number: MFCD00019048
CAS Number: 16433-88-8

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR