2-Chloro-1,4-dibromobenzene ≥99%

Supplier: Matrix Scientific

Synonyms: 2,5-Dibromochlorobenzene, 1,4-Dibromo-2-chlorobenzene, 1-Chloro-2,5-dibromobenzene

004572-25G 004572-100G
101787-504EA 58.46 USD
101787-504 102550-848
2-Chloro-1,4-dibromobenzene ≥99%
2-Chloro-1,4-dibromobenzene

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₃Br₂Cl
MW: 270.35 g/mol
Boiling Pt: 115 °C (20mm)
Melting Pt: 39…41 °C
MDL Number: MFCD00018094
CAS Number: 3460-24-0

Order Now

Specification Test Results

Boiling point 115°C/20mm
Melting point 39-41°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR