2,5-Dibromo-α,α,α-trifluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride

004569-5G 004569-25G
101787-498EA 29.23 USD
101787-498 101787-500
2,5-Dibromo-α,α,α-trifluorotoluene ≥97%
2,5-Dibromo-α,α,α-trifluorotoluene

Hazard code(s): Irritant

Formula: C₇H₃Br₂F₃
MW: 303.9 g/mol
Melting Pt: 47…49 °C
MDL Number: MFCD00013554
CAS Number: 7657-09-2
EINECS: 231-620-1

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR