1,2-Dibromobenzene ≥98%

Supplier: Matrix Scientific

004566-25G 004566-100G
101787-494EA 21.54 USD
101787-494 101787-496
1,2-Dibromobenzene ≥98%
1,2-Dibromobenzene

Hazard code(s): Irritant

Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 224 °C
Melting Pt: 4…6 °C
Density: 1.956 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000057
CAS Number: 583-53-9
EINECS: 209-507-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Boiling point 224°C
Melting point 4-6°C
Density 1.956
ND20 1.611

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR