3,5-Dibromo-4-methoxybenzoic acid

Supplier: Matrix Scientific

004555-5G 004555-25G
101787-466EA 66.15 USD
101787-466 101787-468
3,5-Dibromo-4-methoxybenzoic acid
3,5-Dibromo-4-methoxybenzoic acid

Hazard code(s): Irritant

Formula: C₈H₆Br₂O₃
Melting Pt: 224…226 °C
MDL Number: MFCD00016511
CAS Number: 4073-35-2

Order Now

Specification Test Results

Melting point 224-226°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR