1,2-Dibromo-4,5-difluorobenzene ≥98%

Supplier: Matrix Scientific
Danger

004549-25G 004549-100G
101787-456EA 38.46 USD
101787-456 101787-458
1,2-Dibromo-4,5-difluorobenzene ≥98%
1,2-Dibromo-4,5-difluorobenzene

Hazard code(s): Irritant

Formula: C₆H₂Br₂F₂
MW: 271.89 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 34 °C
MDL Number: MFCD00009890
CAS Number: 64695-78-9
EINECS: 265-021-1
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Melting point 34°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR