1,2-Dibromo-3-chloropropane ≥98%
Supplier: Matrix Scientific
|
Danger
|
Synonyms:
1-Chloro-2,3-dibromopropane
004547-5G
004547-25G
101787-448EA
56.92
USD
101787-448
101787-450
1,2-Dibromo-3-chloropropane ≥98%
1,2-Dibromo-3-chloropropane
Hazard code(s): Toxic
Formula:
ClH₂CBrCHCH₂Br MW: 236.33 g/mol Boiling Pt: 196 °C Melting Pt: 6.1 °C Density: 2.08 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00039365 CAS Number: 96-12-8 EINECS: 202-479-3 UN: 2872 ADR: 6.1 III Merck Index: 12,03068 |
Specification Test Results
Boiling point | 196°C |
Density | 2.08 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...