1,2-Dibromo-1,1-dichloro-2,2-difluoroethane ≥97%
Supplier: Matrix Scientific
004530-1G
004530-5G
101787-400EA
30.77
USD
101787-400
101787-402
1,2-Dibromo-1,1-dichloro-2,2-difluoroethane ≥97%
1,2-Dibromo-1,1-dichloro-2,2-difluoroethane
Hazard code(s): Irritant
Formula:
C₂Br₂Cl₂F₂ Boiling Pt: 138…140 °C Melting Pt: >40 °C |
MDL Number:
MFCD00039319 CAS Number: 558-57-6 |
Specification Test Results
Boiling point | 138-140°C |
Melting point | >40°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...