2-Iodo-5-methylthiophene ≥99% for synthesis

Supplier: Matrix Scientific

004502-1G 004502-5G 004502-25G
101787-338EA 27.69 USD
101787-338 101787-340 101787-342
2-Iodo-5-methylthiophene ≥99% for synthesis
2-Iodo-5-methylthiophene

Hazard code(s): Irritant

Formula: C₅H₅IS
Boiling Pt: 81…83 °C (10mm)
Density: 1.84 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00022492
CAS Number: 16494-36-3

Order Now

Specification Test Results

Boiling point 81-83°C/10m
Density 1.84


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR