4-Iodo-m-toluic acid ≥98%

Supplier: Matrix Scientific

004491-5G 004491-25G
101787-332EA 84.62 USD
101787-332 101787-334
4-Iodo-m-toluic acid ≥98%
4-Iodo-m-toluic acid

Hazard code(s): Light Sensitive

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Melting Pt: 223…225 °C
MDL Number: MFCD00270113
CAS Number: 52107-87-6

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR