3-Iodo-o-toluic acid ≥98%

Supplier: Matrix Scientific

004490-5G 004490-25G
101787-328EA 38.46 USD
101787-328 101787-330
3-Iodo-o-toluic acid ≥98%
3-Iodo-o-toluic acid

Hazard code(s): Light Sensitive

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD00270112
CAS Number: 133232-56-1

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR