6-Iodo-m-toluic acid ≥98%

Supplier: Matrix Scientific

004488-5G 004488-25G 004488-100G
101787-322EA 16.92 USD
101787-322 101787-324 101787-326
6-Iodo-m-toluic acid ≥98%
6-Iodo-m-toluic acid

Hazard code(s): Light Sensitive

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00079765
CAS Number: 52548-14-8

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR