2-Iodo-m-toluic acid ≥98%

Supplier: Matrix Scientific

004487-1G 004487-5G 004487-25G
101787-318EA 32.31 USD
101787-318 101787-320 102550-846
2-Iodo-m-toluic acid ≥98%
2-Iodo-m-toluic acid

Hazard code(s): Light Sensitive

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Melting Pt: 150…153 °C
MDL Number: MFCD00079764
CAS Number: 108078-14-4

Order Now

Specification Test Results

Melting point 150-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR