4-Iodo-o-toluidine ≥98%

Supplier: Matrix Scientific

004486-5G 004486-25G
101787-314EA 16.92 USD
101787-314 101787-316
4-Iodo-o-toluidine ≥98%
4-Iodo-o-toluidine

Hazard code(s): Irritant, Light Sensitive

Formula: C₇H₈IN
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 86…87 °C
MDL Number: MFCD00025299
CAS Number: 13194-68-8
EINECS: 236-154-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 86-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR