(2,3,4-Trimethoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

004466-1G 004466-5G
101787-280EA 133.85 USD
101787-280 101787-282
(2,3,4-Trimethoxyphenyl)acetic acid ≥98%
(2,3,4-Trimethoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₆H₂(OCH₃)₃CH₂COOH
MW: 226.23 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD00016820
CAS Number: 22480-91-7
EINECS: 245-024-4

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR