3-Iodo-4-methoxybenzoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Iodo-p-anisic acid

004457-5G 004457-25G
101787-268EA 36.92 USD
101787-268 101787-270
3-Iodo-4-methoxybenzoic acid ≥98%
3-Iodo-4-methoxybenzoic acid

Hazard code(s): Irritant

Formula: C₈H₇IO₃
MW: 278.05 g/mol
Melting Pt: 245…248 °C
MDL Number: MFCD00017347
CAS Number: 68507-19-7

Order Now

Specification Test Results

Melting point 245-248°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR