5-Chloro-m-xylene ≥98%
Supplier: Matrix Scientific
004445-5G
004445-25G
101787-260EA
80
USD
101787-260
101787-262
5-Chloro-m-xylene ≥98%
5-Chloro-m-xylene
Hazard code(s): Irritant
Formula:
C₈H₉Cl Boiling Pt: 187 °C Density: 1.04 g/cm³ |
MDL Number:
MFCD00060273 CAS Number: 556-97-8 |
Specification Test Results
Boiling point | 187°C |
Density | 1.04 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...