2-Chloro-m-xylene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-1,3-dimethylbenzene, 1-Chloro-2,6-dimethylbenzene

004443-25G 004443-100G
101787-256EA 116.92 USD
101787-256 101787-258
2-Chloro-m-xylene ≥97%
2-Chloro-m-xylene

Hazard code(s): Irritant

Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 182…183 °C
Melting Pt: –35 °C
Density: 1.053 g/cm³ (20 °C)
MDL Number: MFCD00000565
CAS Number: 6781-98-2
EINECS: 229-843-4

Order Now

Specification Test Results

Boiling point 182-183°C
Density 1.06

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR